Orders LinkContactUsLinkComponent
Process wipes, KIMTECH PREP*
Brag-Box, Blue, 310×425 mm, 1 box with 160 sheets
Catalog # 115-1557
Supplier:   KIMBERLY CLARK
Process wipes, KIMTECH PREP*
Catalog # 115-1557
Supplier:  KIMBERLY CLARK
Process wipes, KIMTECH PREP*
Process wipes, KIMTECH PREP*
Brag-Box, Blue, 310×425 mm, 1 box with 160 sheets
Price
124,00 €
Log in for availability
1 * 160 ST
Product Details & Documents
Strong, low-lint wipes for repeat use cleaning with chemicals and solvents. Perfect for printing, pre and post-paint applications, glass and appliance manufacturing, engine maintenance.
  • PP, reusable
  • Great for wiping oils and grease
  • Acid, base, and solvent resistant
  • Roll 115-2222 is compatible with dispensers 115-0045, 115-1533 and 115-2038
Colour: Blue
Specifications
  • Packed:
    1 box with 160 sheets
  • Colour:
    Blue
  • Description:
    Brag-Box
  • L×W:
    310×425 mm