Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PRONTOS 300-5-C18 ACE-EPS 250X4.6MM 1 * 1 ST
Catalog # BISC2546K18APS050
Supplier:
BISCHOFF ANALYSETECHNIK
Expand image
PRONTOS 300-5-C18 ACE-EPS 250X4.6MM 1 * 1 ST
Catalog # BISC2546K18APS050
Supplier:
BISCHOFF ANALYSETECHNIK