Orders LinkContactUsLinkComponent
Software for Julabo units
Software for Julabo units
EasyTemp Professional software, controlling up to 24 Julabo units with RS232 interface
Catalog # 461-0063
Supplier:   JULABO GmbH
Software for Julabo units
Catalog # 461-0063
Supplier:  JULABO GmbH
Software for Julabo units
Software for Julabo units
EasyTemp Professional software, controlling up to 24 Julabo units with RS232 interface
Price
2 500,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
A range of software that allows programming, viewing and recording of temperature and time dependent processes in combination with laboratory instruments. Comprehensive communication and networking possibilities via RS232/RS485, USB (with optional converter) and Profibus assist with laboratory automation.
  • Start/stop functions and set point input with graphical zoom functions
  • Indication of valid actual, heating and cooling capacity
  • Repeat memorised profiles
  • Display of up to four curves in one diagram
  • Ramp function of up to 100 steps
EasyTemp software: Can be downloaded free of charge from the Julabo website. It is ideal for use with Julabo units with RS232 interface, enabling control of a single unit. Recording of measured values in ASCII format.
EasyTemp Professional software: Enables control of up to 24 Julabo units with RS232 (USB optional) interface. Recording of measured values in ASCII and Excel format. Up to 4 definable scales, user can enter formulae such as averaging, differences between measured values, and text comments can be displayed in the graph.
Programming and recording of individual profiles for each unit, with graphics display of up to 16 curves in one diagram
Reproduces the important values in a graph
Separate window for controlling each unit, one button can start units simultaneously
Profiles can be changed whilst running
Comprehensive ramp programming with up to 1000 steps per profile, for each unit
Options for EasyTemp Professional include control of laboratory instruments of different makes, e.g. laboratory stirrers, magnetic stirrers, dosing pumps, pH measurements. Driver for Devicenet, Profibus, Ethernet and RS485 interfaces are also possible, please contact your local Avantor sales office.
Specifications
  • Description:
    EasyTemp Professional software
  • For:
    controlling up to 24 Julabo units with RS232 interface