Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Sucrose

D-(+)-Sucrose

Glass bottle for solids, 500 g
Catalog # BMOL21986
D-(+)-Sucrose
Catalog # BMOL21986
CAS Number:  57-50-1
D-(+)-Sucrose
D-(+)-Sucrose

Glass bottle for solids, 500 g

CAS Number:  57-50-1
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-334-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.