Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Both ends open, 100 mm, 1,8/2,0 mm
Catalog # 612-4867
Supplier:   MURRAY S
Capillary tubes
Catalog # 612-4867
Supplier:  MURRAY S
Capillary tubes
Capillary tubes
Both ends open, 100 mm, 1,8/2,0 mm
Price
116,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Soda glass.
  • For melting point determinations
Specifications
  • Ext.Ø:
    1,8/2,0 mm
  • Description:
    Both ends open
  • Length:
    100 mm