Capillary tubes
Both ends open, 75 mm, 1,8/2,0 mm
Catalog # 612-4866
Supplier: MURRAY S
Jump to:
Product Details & Documents
Soda glass.
- For melting point determinations
Specifications
- Ext.Ø:1,8/2,0 mm
- Description:Both ends open
- Length:75 mm