Acetone ≥99%
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
Acetone ≥99%
Catalog # L10407.AU
Supplier: Thermo Fisher Scientific
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Refractive Index: 1.3590 Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:14,66
- Boiling Pt:56 °C
- Melting Pt:–94 °C
- Density:0.791
- Flash Pt:–17 °C (1 °F)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Cat. No.:L10407.AU
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



