Calibration weights, cylindrical, OIML, class E2
50 g, 0,10 ± mg, Cylindrical test weight
Catalog # SARBYCW4528-02
Supplier: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Catalog # SARBYCW4528-02
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifications
- Weight:50 g
- Seq:20
- Tolerance:0,10 ± mg
- Type:Cylindrical test weight