First-aid kits
Industrial first aid kit, 1 - 20 people, supplied in shatterproof ABS case
Artikelnummer 118-0121
Hersteller: SAFETY FIRST AID GROUP
Jump to:
Product Details & Documents
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Delivery information: All kits are supplied with a first aid guidance leaflet.
Specifications
- Description:Industrial first aid kit, 1 - 20 people, supplied in shatterproof ABS case
- Cat. No.:118-0121
- Contents:120 × Waterproof plasters, sterile
40 × individually wrapped wipes
12 × blue plastic fingerstalls (assorted)
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 × skin closure adhesives, sterile
10 × eyewash, sterile
2 × hygienic resuscitation kit
8 × Primapore wound dressings (80×80 mm)
4 × Primapore wound dressings (80×80 mm)
1 × emergency foil blanket
1 × dressing scissors
100 disposable examination gloves (pairs)
1 wall bracket - Model:Industrial (1 - 20 persons)