Orders LinkContactUsLinkComponent
Draining racks, Azlon®
Draining racks, Azlon®
Set of 11 narrow pegs
Catalog # AZLOWRP002P
Supplier:   SciLabware
Draining racks, Azlon®
Catalog # AZLOWRP002P
Supplier:  SciLabware
Draining racks, Azlon®
Draining racks, Azlon®
Set of 11 narrow pegs
Price
54,90 €
Log in for availability
1 * 1 SET
Product Details & Documents
Easy to assemble, lightweight polystyrene draining racks designed to be wall mounted.
  • Pegs slot into place on the back board and are spaced to allow various sizes of lab ware to fit on to the rack
  • Water is collected in the trough at the base of the rack and drains away via a rubber tube which can be directed into a nearby sink or container
Ø/peg: 16 mm
Delivery information: Supplied with wall mounting accessories.
Specifications
  • Description:
    Set of 11 narrow pegs
  • Seq:
    02