Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cleaning brushes for samplers

Nylon, 15 - 30 mm, Wire, 1100 mm
Catalog # 300-0492
Supplier:   Sampling Systems
Cleaning brushes for samplers
Catalog # 300-0492
Supplier:  Sampling Systems
Cleaning brushes for samplers
Cleaning brushes for samplers

Nylon, 15 - 30 mm, Wire, 1100 mm

Price
55,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.

  • Available in a range of lengths and diameters

Documents

Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    15 - 30 mm
  • Handle material:
    Wire
  • Seq:
    06
  • Total length:
    1100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.