4-Fluorophenol ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 371-41-5 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 68°(154°F) Odour: Phenol-like Boiling Point: 185-188° Beilstein Reference: 1362752 UN No.: 2923 MDL No.: MFCD00002316
Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
- CAS Number:371-41-5
- Formula:C₆H₅FO
- MDL Number:MFCD00002316
- Molecular Weight:112.1 g/mol
- Boiling Pt:185…188 °C
- Melting Pt:44…50 °C
- Density:1.22
- Flash Pt:68 °C (154 °F)
- Storage Temperature:Ambient
- EINECS:206-736-0
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



