Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall
100 mm, 16 mm
  212-7108
 :   MURRAY S
Test tubes without rim, medium wall
  212-7108
 :  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall
100 mm, 16 mm
97,50 €
1 * 100 ST
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
 : ISO 4142:2002
  • Ext.Ø:
    16 mm
  • Length:
    100 mm