Order Entry
Germany
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rod, PP, 3 mm, 120 mm
Catalog # SARS81.970
Marketsource
Supplier:   SARSTEDT
Stirring rods
Catalog # SARS81.970
Supplier:  SARSTEDT
Marketsource
Stirring rods
Stirring rods

Stirring rod, PP, 3 mm, 120 mm

Price
128,00 €
Log in for availability
1 * 10.000 ST
Product Details & Documents

Stirring rods.

Specifications
  • Ø:
    3 mm
  • Description:
    Stirring rod
  • Length:
    120 mm
  • Material:
    PP
  • Seq:
    01