Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

10,01, Sachet, 25×20 ml, 25
Catalog # 662-4005
Supplier:   Hanna
Buffer solutions
Catalog # 662-4005
Supplier:  Hanna
CAS Number:  
Buffer solutions
Buffer solutions

10,01, Sachet, 25×20 ml, 25

CAS Number:  
Price
63,50 €
Log in for availability
1 * 25 ST
Product Details & Documents

Solutions in sachets ensure optimum precision. The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH meter that has been calibrated according to the NIST standards.

  • Accurate
  • Disposable
  • No danger of contamination

Delivery information: The buffers in sachets are delivered with a certificate of conformity.

Documents

Specifications

Specifications

  • Pack type:
    Sachet
  • Pk:
    25
  • Pk Info:
    25×20 ml
  • pH value:
    10,01

Specifications