Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions

7,01, Sachet, 25×20 ml, 25
Catalog # 662-4004
Supplier:   Hanna
Buffer solutions
Catalog # 662-4004
Supplier:  Hanna
CAS Number:  
Buffer solutions
Buffer solutions

7,01, Sachet, 25×20 ml, 25

CAS Number:  
Price
63,50 €
Log in for availability
1 * 25 ST
Product Details & Documents

Solutions in sachets ensure optimum precision. The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH meter that has been calibrated according to the NIST standards.

  • Accurate
  • Disposable
  • No danger of contamination

 : The buffers in sachets are delivered with a certificate of conformity.

 

Specifications

Specifications

  • Pack type:
    Sachet
  • Pk:
    25
  • Pk Info:
    25×20 ml
  • pH value:
    7,01
  • Cat. No.:
    662-4004

Specifications