Septa
Septa, 12 mm, 2 mm
Catalog # METR6.1448.010
Supplier: METROHM
Septa
Septa, 12 mm, 2 mm
Jump to:
Product Details & Documents
These septa are made from silicone rubber.
Specifications
- Ø:12 mm
- Description:Septa
- Seq:01
- Thickness:2 mm
Nickel aluminium alloy, powder for production of Raney-nickel, for synthesis, Sigma-Aldrich®
#: 8.06749.0005
Supplier: Merck
Silver nitrate standardised solution 0.1 N in aqueous solution
#: 35601.K2
Supplier: Thermo Fisher Scientific
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
#: EXL-1211-1546U
Supplier: Avantor
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
#: 85930.150
Supplier: VWR Chemicals
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
#: 34800-6X1L
Supplier: Honeywell Chemicals
Potassium bromate ≥99.8%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
#: 1.04912.0100
Supplier: Merck
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
#: 663-0014
Supplier: Mettler - Toledo
BLAUBRAND® ETERNA Graduated Pipettes, ISO, Type 3, Class AS, Total Delivery (One Batch)
#: BRND2.28406
Supplier: Brand



