Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Spoon spatulas, broad shape

150 mm, 20 mm, 2 ml
  231-0123
Spoon spatulas, broad shape
  231-0123
Spoon spatulas, broad shape
Spoon spatulas, broad shape

150 mm, 20 mm, 2 ml

 
103,00 €
   
1 * 10 ST
 

Stainless steel.

  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
  • Non magnetic, bright finish
 
  • Capacity:
    2 ml
  • Length:
    150 mm
  • Seq:
    02
  • Spoon size:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.