Order Entry
Germany
Orders LinkContactUsLinkComponent
Septa

Septa

Septa, 16 mm, 2 mm
Catalog # METR6.1448.020
Supplier:   METROHM
Septa
Catalog # METR6.1448.020
Supplier:  METROHM
Septa
Septa

Septa, 16 mm, 2 mm

Price
31,10 €
Log in for availability
1 * 5 ST
Product Details & Documents

These septa are made from silicone rubber.

Specifications
  • Ø:
    16 mm
  • Description:
    Septa
  • Seq:
    02
  • Thickness:
    2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.