Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Chlorophenyl)propionic acid ≥98%

3-(2-Chlorophenyl)propionic acid ≥98%

Glass bottle for solids, 5 g
Catalog # L15306.06
Irritant
3-(2-Chlorophenyl)propionic acid ≥98%
Catalog # L15306.06
CAS Number:  1643-28-3
3-(2-Chlorophenyl)propionic acid ≥98%
3-(2-Chlorophenyl)propionic acid ≥98%

Glass bottle for solids, 5 g

CAS Number:  1643-28-3
Price
205,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 1643-28-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 94-97° Boiling Point: 171°/10mm MDL No.: MFCD00016547

Synonyms:  3-(2-Chlorophenyl)propanoic acid
  • CAS Number:1643-28-3
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00016547
  • Molecular Weight:184.62 g/mol
  • Boiling Pt:171 °C (10 mmHg)
  • Melting Pt:94…97 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.