Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Chlorophenyl)propionic acid ≥98%

3-(2-Chlorophenyl)propionic acid ≥98%

Glass bottle for solids, 1 g
Catalog # L15306.03
Irritant
3-(2-Chlorophenyl)propionic acid ≥98%
Catalog # L15306.03
CAS Number:  1643-28-3
3-(2-Chlorophenyl)propionic acid ≥98%
3-(2-Chlorophenyl)propionic acid ≥98%

Glass bottle for solids, 1 g

CAS Number:  1643-28-3
Price
56,20 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 1643-28-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 94-97° Boiling Point: 171°/10mm MDL No.: MFCD00016547

Synonyms:  3-(2-Chlorophenyl)propanoic acid
  • CAS Number:1643-28-3
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00016547
  • Molecular Weight:184.62 g/mol
  • Boiling Pt:171 °C (10 mmHg)
  • Melting Pt:94…97 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications