Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

Glass bottle, 500 mL
Catalog # L15029.AP
Polysorbate 20
Catalog # L15029.AP
CAS Number:  9005-64-5
Polysorbate 20
Polysorbate 20

Glass bottle, 500 mL

CAS Number:  9005-64-5
Price
41,70 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 9005-64-5 Molecular Formula: C58H114O26 Flash Point: 110°(230°F) Refractive Index: 1.4700 Boiling Point: 100° MDL No.: MFCD00165986

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Density:1.105
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.