Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Polysorbate 20

Glass bottle, 100 mL
Catalog # L15029.AE
Polysorbate 20
Catalog # L15029.AE
CAS Number:  9005-64-5
Polysorbate 20
Polysorbate 20

Glass bottle, 100 mL

CAS Number:  9005-64-5
Price
25,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 9005-64-5 Molecular Formula: C58H114O26 Flash Point: 110°(230°F) Refractive Index: 1.4700 Boiling Point: 100° MDL No.: MFCD00165986

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Density:1.105
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications