Polysorbate 80
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:14,7582
- Boiling Pt:>100 °C
- Melting Pt:100 °C
- Density:1.064
- Flash Pt:>149 °C (300 °F)
- Storage Temperature:Ambient
- EINECS:500-019-9
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



