Order Entry
Germany
Orders LinkContactUsLinkComponent
Cover slips for microscope slides

Cover slips for microscope slides

Cover slip for microscope slide, - mm, 0,15 - 0,25 mm, 25,4 mm, Quartz
Catalog # 43211.KJ
Cover slips for microscope slides
Catalog # 43211.KJ
Cover slips for microscope slides

Cover slip for microscope slide, - mm, 0,15 - 0,25 mm, 25,4 mm, Quartz

Price
238,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

These cover slips made of fused quartz, have many properties, due to the fusing of naturally occuring crystalline silica.

  • High chemical purity and corrosion resistance
  • Low thermal expansion coefficient
  • Extreme hardness

Excellent electrical insulation and optical transmission from ultraviolet to infrared are suitable for analytical purposes.

Specifications
  • Ø:
    25,4 mm
  • Description:
    Cover slip for microscope slide
  • L×W:
    - mm
  • Material:
    Quartz
  • Seq:
    04
  • Thickness:
    0,15 - 0,25 mm