Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrene ≥98%

Pyrene ≥98%

Glass bottle for solids, 25 g
Catalog # L08162.14
Pyrene ≥98%
Catalog # L08162.14
CAS Number:  129-00-0
Pyrene ≥98%
Pyrene ≥98%

Glass bottle for solids, 25 g

CAS Number:  129-00-0
Price
35,60 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 129-00-0 Molecular Formula: C16H10 Formula Weight: 202.26 Flash Point: 208°(406°F) Melting Point: 148-152° Boiling Point: 393° Beilstein Reference: 1307225 Merck Reference: 14,7963 MDL No.: MFCD00004136

  • CAS Number:129-00-0
  • Formula:C₁₆H₁₀
  • MDL Number:MFCD00004136
  • Molecular Weight:202.26 g/mol
  • Merck Index:14,7963
  • Boiling Pt:393 °C
  • Melting Pt:148…152 °C
  • Density:1.271
  • Flash Pt:208 °C (406 °F)
  • Storage Temperature:Ambient
  • EINECS:204-927-3
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.