Bromophenol blue in aqueous solution
Plastic bottle, 250 mL
Catalog # LOVI451622
Supplier: Lovibond
Wechseln zu:
Product Details & Documents
Synonyme: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS-Nummer:115-39-9
- Formel::C₁₉H₁₀Br₄O₅S
- MDL-Nummer:MFCD00005875
- Molekulargewicht:669.97 g/mol
- Lagertemperatur:Ambient
- EINECS:204-086-2
Spezifikationen:
Spezifikationen:
- Pack type:Plastic bottle
- Pk:250 mL
Spezifikationen:
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



