Orders LinkContactUsLinkComponent
Desiccators, PYREX®
Rotaflo® stopcock with 24/29 joint for vacuum tight seal
Catalog # BIBB1612/03
Supplier:   SciLabware
Desiccators, PYREX®
Catalog # BIBB1612/03
Supplier:  SciLabware
Desiccators, PYREX®
Desiccators, PYREX®
Rotaflo® stopcock with 24/29 joint for vacuum tight seal
Price
57,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Ideal for vacuum and non vacuum applications.
  • Ground glass flanges
  • Borosilicate glass base and lid
  • Plates positioned on an internal ledge within the base
Specifications
  • Description:
    Rotaflo® stopcock with 24/29 joint for vacuum tight seal
  • Seq:
    065