ContactUsLinkComponent
Clip, rotulex joint metal clip, spherical joint approx. bore 40 mm to fit rm:rf64 max outer diameter shank 45 mm spherical diameter 64 mm. 1 * 5 items
Catalog # BIBBRC64
Supplier:  SciLabware
undefined
Clip, rotulex joint metal clip, spherical joint approx. bore 40 mm to fit rm:rf64 max outer diameter shank 45 mm spherical diameter 64 mm. 1 * 5 items
Catalog # BIBBRC64
Supplier:  SciLabware
Frequently Bought Together