Order Entry
Germany
Orders LinkContactUsLinkComponent
Scoops, Azlon®

Scoops, Azlon®

Scoops, chemical resistand and food safe, 1500 ml, 405 mm, HDPE
Catalog # AZLOSWL040
Supplier:   SciLabware
Scoops, Azlon®
Catalog # AZLOSWL040
Supplier:  SciLabware
Scoops, Azlon®
Scoops, Azlon®

Scoops, chemical resistand and food safe, 1500 ml, 405 mm, HDPE

Price
27,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sturdy scoops made from natural rigid PE.

  • Lightweight
  • Good chemical resistance
  • Food safe
Specifications
  • Capacity:
    1500 ml
  • Description:
    Scoops, chemical resistand and food safe
  • Length:
    405 mm
  • Material:
    HDPE
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.