Orders LinkContactUsLinkComponent
Swabs tubes
Swabs tubes
Plain swabs (bulk), Wood/cotton, +
Catalog # GREISWA5_1000
Supplier:   Greiner Bio-One
Swabs tubes
Catalog # GREISWA5_1000
Supplier:  Greiner Bio-One
Swabs tubes
Plain swabs (bulk), Wood/cotton, +
Price
32,90 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Swab in tubes suitable for the collection and transport of non-human bacteriological, serological, and cytological samples.
  • Sterilised by irradiation
  • Swab tubes made of polystyrene
Useful for hygienic controls in veterinary research as well as in environmental sampling.
Packaging: Individually wrapped.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Sterile:
    +
  • Description:
    Plain swabs (bulk)
  • Material:
    Wood/cotton