Trypsin
Trypsin-EDTA 10X, 100 mL
Supplier: Biowest
Jump to:
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Documents
Specifications
Specifications
- Pk:100 mL
- Description:Trypsin-EDTA 10X