Orders LinkContactUsLinkComponent
Trypsin
Trypsin
Trypsin-EDTA 1X, with sodium chloride, lyophilised, 1 g
Catalog # P0940-1GR
Supplier:   Biowest
Trypsin
Catalog # P0940-1GR
Supplier:  Biowest
CAS Number:  
Trypsin
Trypsin
Trypsin-EDTA 1X, with sodium chloride, lyophilised, 1 g
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifications
Specifications
  • Pk:
    1 g
  • Description:
    Trypsin-EDTA 1X, with sodium chloride, lyophilised