Orders LinkContactUsLinkComponent
Trypsin
Trypsin
Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 500 mL
Catalog # L0940-500
Supplier:   Biowest
Trypsin
Catalog # L0940-500
Supplier:  Biowest
CAS Number:  
Trypsin
Trypsin
Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 500 mL
CAS Number:  
Price
43,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red
Specifications