Orders LinkContactUsLinkComponent
Trypsin
Trypsin
Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 100 mL
Artikelnummer L0940-100
Hersteller:   Biowest
Trypsin
Artikelnummer L0940-100
Hersteller:  Biowest
CAS-Nummer:  
Trypsin
Trypsin
Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 100 mL
CAS-Nummer:  
Preis
12,70 €
Log in for availability
1 * 100 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifications
Specifications
  • Pk:
    100 mL
  • Description:
    Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red