Order Entry
Germany
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

Trypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol red, 100 ml
Catalog # L0930-100
Supplier:   Biowest
Trypsin
Catalog # L0930-100
Supplier:  Biowest
CAS Number:  
Trypsin
Trypsin

Trypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol red, 100 ml

CAS Number:  
Price
12,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Trypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol red

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.