Buffer, PBS solution 10X concentrate (phosphate buffered saline)
PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 500 mL
Catalog # TREV4870-500
Marketsource
Supplier: TREVIGEN
Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # TREV4870-500
Supplier: TREVIGEN
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:PBS buffer solution 10X concentrate (phosphate buffered saline)



