Orders LinkContactUsLinkComponent
Flame photometric detectors
FPD ignitor replacement kit
  HEWL19256-60800
 :   Agilent
Flame photometric detectors
  HEWL19256-60800
 :  Agilent
Flame photometric detectors
Flame photometric detectors
FPD ignitor replacement kit
154,00 €
1 * 1 KIT
FPD is designed with minimum detectable levels of 3,6 pg/s for sulfur and 60 fg/s for phosphorus.
 : Upgrade kits are available and can be ordered separately.
  • Description:
    FPD ignitor replacement kit
  • Seq:
    03