Orders LinkContactUsLinkComponent
Flame photometric detectors | Avantor
Flame photometric detectors
O-ring
Catalog # HEWL0905-1610
Supplier:   Agilent
Flame photometric detectors
Catalog # HEWL0905-1610
Supplier:  Agilent
Flame photometric detectors
Flame photometric detectors
O-ring
Wechseln zu:
Price
82,00 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents
FPD is designed with minimum detectable levels of 3,6 pg/s for sulfur and 60 fg/s for phosphorus.
Bestellinformationen: Upgrade kits are available and can be ordered separately.
Spezifikationen:
  • Description:
    O-ring
  • Seq:
    01