Order Entry
Germany
Orders LinkContactUsLinkComponent
Sample bottles without push-on Closure

Sample bottles without push-on Closure

25 ml, 51 mm, 30 mm
Catalog # 215-3234
Supplier:   GRAVIS
Sample bottles without push-on Closure
Catalog # 215-3234
Supplier:  GRAVIS
Sample bottles without push-on Closure
Sample bottles without push-on Closure

25 ml, 51 mm, 30 mm

Price
171,00 €
Log in for availability
1 * 210 ST
Product Details & Documents

Chemistry glass, white or amber, without push-on cap.

  • Amber glass to protect light-sensitive materials

Ordering information: Push-on PE caps are available separately.

Documents

Specifications
  • Kapazität:
    25 ml
  • Höhe:
    51 mm
  • Ø Hals:
    30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.