4-Hydroxybenzhydrazide
Glass bottle for solids, 10 g
Catalog # FLUO018715-10G
Marketsource
Supplier: FLUOROCHEM
4-Hydroxybenzhydrazide
Catalog # FLUO018715-10G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 5351-23-5
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
- CAS Number:5351-23-5
- Formula:C₇H₈N₂O₂
- MDL Number:MFCD00007605
- Molecular Weight:152.15 g/mol
- Boiling Pt:370 °C (1013 hPa)
- Melting Pt:266…269 °C
- Storage Temperature:Ambient
- EINECS:226-326-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



