Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3-(2-Bromophenyl)propionic acid

Glass bottle for solids, 5 g
Catalog # FLUO018310-5G
Marketsource
Supplier:   FLUOROCHEM
3-(2-Bromophenyl)propionic acid
Catalog # FLUO018310-5G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  15115-58-9
3-(2-Bromophenyl)propionic acid

Glass bottle for solids, 5 g

CAS Number:  15115-58-9
Price
47,20 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:15115-58-9
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310791
  • Molecular Weight:229.07 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications