Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Bromo-2'-hydroxyacetophenone

5'-Bromo-2'-hydroxyacetophenone

25 g
Catalog # FLUO012070-25G
Marketsource
Supplier:   FLUOROCHEM
5'-Bromo-2'-hydroxyacetophenone
Catalog # FLUO012070-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  1450-75-5
5'-Bromo-2'-hydroxyacetophenone

25 g

CAS Number:  1450-75-5
Price
59,70 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
  • CAS Number:1450-75-5
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00191850
  • Molecular Weight:215.05
Specifications

Specifications

  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.