5'-Bromo-2'-hydroxypropiophenone
1
Catalog # CHMPFL12088.POR
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one
- CAS Number:17764-93-1
- Formula:C₉H₉BrO₂
- MDL Number:MFCD00218577
- Molecular Weight:229.0706 g/mol
Specifications
Specifications
- Pk:1



