Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-4-nitrobenzonitrile

2-Hydroxy-4-nitrobenzonitrile

1
Catalog # CHMPFL12255.POR
Supplier:   CHEMPUR
2-Hydroxy-4-nitrobenzonitrile
Catalog # CHMPFL12255.POR
Supplier:  CHEMPUR
CAS Number:  39835-14-8
2-Hydroxy-4-nitrobenzonitrile

1

CAS Number:  39835-14-8
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
  • CAS Number:39835-14-8
  • Formula:C₇H₄N₂O₃
  • MDL Number:MFCD02258875
  • Molecular Weight:164.1183 g/mol
Specifications

Specifications

  • Pk:
    1

Specifications