3'-Nitroformanilide
Glass bottle for solids, 100 g
Catalog # FLUO011202-100G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: 3-Nitroformanilide
- CAS Number:102-38-5
- Formula:C₇H₆N₂O₃
- MDL Number:MFCD00017014
- Molecular Weight:166.14 g/mol
- Melting Pt:134 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



