Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Ethoxynicotinic acid

2-Ethoxynicotinic acid

Glass bottle for solids, 100 g
Catalog # FLUO009781-100G
Marketsource
Supplier:   FLUOROCHEM
2-Ethoxynicotinic acid
Catalog # FLUO009781-100G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  35969-54-1
2-Ethoxynicotinic acid

Glass bottle for solids, 100 g

CAS Number:  35969-54-1
Price
737,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:35969-54-1
  • Formula:C₈H₉NO₃
  • MDL Number:MFCD00128788
  • Molecular Weight:167.16 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.