2-Ethoxynicotinic acid
Glass bottle for solids, 25 g
Catalog # FLUO009781-25G
Marketsource
Supplier: FLUOROCHEM
2-Ethoxynicotinic acid
Catalog # FLUO009781-25G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 35969-54-1
Jump to:
Product Details & Documents
- CAS Number:35969-54-1
- Formula:C₈H₉NO₃
- MDL Number:MFCD00128788
- Molecular Weight:167.16 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



