Strontium fluoride
Glass bottle for solids, 500 g
Catalog # FLUO007296-500G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:7783-48-4
- Formula:F₂Sr
- MDL Number:MFCD00011251
- Molecular Weight:125.62 g/mol
- Merck Index:12,09002
- Boiling Pt:2460 °C (1013 hPa)
- Melting Pt:1477 °C
- Density:4.24 g/cm³ (20 °C)
- EINECS:232-000-3
- UN:3288
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



