Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Chloro-3-fluorophenol

4-Chloro-3-fluorophenol

Glass bottle for solids, 25 g
Catalog # FLUO004459-25G
Marketsource
Supplier:   FLUOROCHEM
4-Chloro-3-fluorophenol
Catalog # FLUO004459-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  348-60-7
4-Chloro-3-fluorophenol

Glass bottle for solids, 25 g

CAS Number:  348-60-7
Price
88,20 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:348-60-7
  • Formula:C₆H₄ClFO
  • MDL Number:MFCD00042583
  • Molecular Weight:146.55 g/mol
  • Boiling Pt:163 °C (1013 hPa)
  • Melting Pt:49…53 °C
  • UN:3261
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications